Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MT-CO2 Rabbit anti-Human, DyLight 405, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB102747 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB102747 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB102747 Supplier Novus Biologicals Supplier No. NBP297593V

Rabbit Polyclonal Antibody

MT-CO2 Polyclonal specifically detects MT-CO2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MT-CO2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias COII, COX2, COXII, Cytochrome C Oxidase II, Cytochrome C Oxidase Polypeptide II, Cytochrome C Oxidase Subunit II, EC 1.9.3.1, Mitochondrially Encoded Cytochrome C Oxidase II, MTCO2
Gene Symbols COX2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MT-CO2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4513
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.