Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTA2 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB003669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003669 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003669 Supplier Novus Biologicals Supplier No. NB10056483V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTA2 Polyclonal specifically detects MTA2 in Human, Primate samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen MTA2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp686F2281, metastasis associated 1 family, member 2, metastasis -associated gene 1-like 1, metastasis associated gene family, member 2, Metastasis-associated 1-like 1, metastasis-associated protein 2, metastasis-associated protein MTA2, MTA1-L1 protein, MTA1L1MTA1-L1, p53 target protein in deacetylase complex, PID
Gene Symbols MTA2
Host Species Rabbit
Immunogen A portion of amino acids 650-700 of human MTA2 was used as the immunogen (NP_004730).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Target Species Human, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.