Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTA3 Antibody
SDP

Catalog No. NB1002306 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002306 Supplier Novus Biologicals Supplier No. NB1002306

Rabbit Polyclonal Antibody has been used in 5 publications

MTA3 Polyclonal specifically detects MTA3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen MTA3
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, Immunohistochemistry 1:500- 1:2000, Immunoprecipitation 2-10 ug/mg of lysate, Immunohistochemistry-Paraffin 1:500- 1:2000
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. Q9BTC8
Gene Alias member 3, metastasis associated 1 family, member 3, metastasis-associated protein MTA3
Gene Symbols MTA3
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 400 and 450 of human metastasis associated 3 using the numbering given in entry Swiss-Prot entry Q9BTC8 (GeneID 57504)
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Breast Cancer
Primary or Secondary Primary
Gene ID (Entrez) 57504
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.