Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTAC2D1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219425 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219425 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219425 Supplier Novus Biologicals Supplier No. NBP219425
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTAC2D1 Polyclonal specifically detects MTAC2D1 in Human samples. It is validated for Western Blot.

Specifications

Antigen MTAC2D1
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Alias C2 calcium-dependent domain containing 1, C2CD1, chromosome 14 open reading frame 47, FLJ36557, membrane targeting (tandem) C2 domain containing 1, Membrane targeting tandem C2 domain-containing protein 1, MTAC2D1, Tac2-NC14orf47, tandem C2 domains nuclear protein, tandem C2 domains, nuclear, Tandem C2 protein in nucleus
Gene Symbols TC2N
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human TC2N. The exact sequence is proprietary.
Molecular Weight of Antigen 55 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123036
Test Specificity Human. Species reactivity based on sequence identity: Bovine (87%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.