Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTAP Antibody (rMTAP/1813), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB383328 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383328 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383328 Supplier Novus Biologicals Supplier No. NBP308964AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MTAP Monoclonal antibody specifically detects MTAP in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen MTAP
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rMTAP/1813
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias 5'-methylthioadenosine phosphorylase, c86fus, EC 2.4.2.28, methylthioadenosine phosphorylase, MSAPMeSAdo phosphorylase, MTA phosphorylase, MTAPase, S-methyl-5'-thioadenosine phosphorylase
Host Species Mouse
Immunogen Recombinant human MTAP protein fragment (aa97-196) (exact sequence is proprietary) (Uniprot: Q13126)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4507
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.