Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTBP Antibody (2C1E11), HRP, Novus Biologicals™
SDP

Catalog No. NBP250517H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250517H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250517H Supplier Novus Biologicals Supplier No. NBP250517H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MTBP Monoclonal specifically detects MTBP in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen MTBP
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 2C1E11
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias hMTBP, MDM2 (mouse double minute 2)-binding protein, 104kD, Mdm2, transformed 3T3 cell double minute 2, p53 binding protein (mouse) bindingprotein, 104kDa, mdm2-binding protein, MDM2BP
Gene Symbols MTBP
Host Species Mouse
Immunogen Recombinant full-length human MTBP
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 27085
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.