Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTCH2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen MTCH2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP154791
SDP
View Documents
Novus Biologicals
NBP154791
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

MTCH2 Polyclonal specifically detects MTCH2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

MTCH2
Polyclonal
Rabbit
Q9Y6C9
23788
Synthetic peptides corresponding to MTCH2(mitochondrial carrier homolog 2 (C. elegans)) The peptide sequence was selected from the N terminal of MTCH2. Peptide sequence TIGRNIFGRQVCQLPGLFSYAQHIASIDGRRGLFTGLTPRLCSGVLGTVV.
Primary
Western Blot
Unconjugated
RUO
2310034D24Rik, Met-induced mitochondrial protein, MIMP, mitochondrial carrier 2, mitochondrial carrier homolog 2, mitochondrial carrier homolog 2 (C. elegans)
MTCH2
IgG
33 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.