Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTCO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | MTCO2 |
|---|---|
| Concentration | 0.37 mg/mL |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5966
![]()
|
Proteintech Group Inc
550701AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody is specific to COX2.
Cytochrome c oxidase is the fourth complex in the respiratory chain and is responsible for catalyzing the conversion of O2 to H2O. Subunit 2 of the cytochrome c oxidase complex combines with two other subunits (1 and 3) to form a core protein structure that performs many functions of the enzyme. The metallic copper center of this subunit transfers electrons to the heme center of subunit 1, which results in the movement of electrons from cytochrome c to the heme A3 and copper B metallic center of complex IV. Defects in subunit 2 of cytochrome c oxidase can result in COX deficiency, which causes a wide range of symptoms from local myopathy to multiple system pathologies that begin between infancy and adulthood. Abnormalities in this subunit are also associated with tumor development.Specifications
| MTCO2 | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P00403, P00405 | |
| 17709, 26198, 4513 | |
| Peptide | |
| Primary | |
| -20°C | |
| COX2 |
| 0.37 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| COII, COX2, COXII, cytochrome c oxidase II, Cytochrome c oxidase subunit 2, MT CO2, MTCO2 | |
| COX2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title