Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTERFD1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB164428 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164428 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164428 Supplier Novus Biologicals Supplier No. NBP281740AF488
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MTERFD1 Polyclonal antibody specifically detects MTERFD1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen MTERFD1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias CGI-12, FLJ10939, Mitochondrial transcription termination factor 3, MTERF domain containing 1, mTERF domain-containing protein 1, mitochondrial, mTERF3
Host Species Rabbit
Immunogen MTERFD1 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human MTERFD1 . The immunogen is located within the last 50 amino acids of MTERFD1. Amino Acid Squence: LGRAQYDPAKPNYISLD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cell Biology, Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 51001
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.