Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 8 publications
$416.50 - $694.00
Specifications
Antigen | MTF1 |
---|---|
Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin Reported in scientific literature (PMID: 24529376)., Immunohistochemistry-Frozen Reported in scientific literature (PMID 24529376 ). |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
MTF1 Polyclonal specifically detects MTF1 in Human, Mouse, Bovine samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
MTF1 | |
Polyclonal | |
Rabbit | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
4520 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:EHSPDNNIIYFEAEEDELTPDDKMLRFVDKNGLVPSSSGTVYDRTTVLIEQDPGTLEDEDDDGQCG | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin Reported in scientific literature (PMID: 24529376)., Immunohistochemistry-Frozen Reported in scientific literature (PMID 24529376 ). | |
Unconjugated | |
RUO | |
metal regulatory transcription factor 1, metal-regulatory transcription factor 1, metal-responsive transcription factor 1, MGC23036, MRE-binding transcription factor, MRE-binding transcription factor-1, MTF-1, Transcription factor MTF-1, zinc regulatory factor, ZRF | |
MTF1 | |
IgG | |
Affinity Purified | |
Specificity of human MTF1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title