Learn More
Description
Specifically detects MTF2 in Human, Mouse samples, and it is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | MTF2 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 |
| Formulation | PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal |
| Gene Alias | metal response element binding transcription factor 2, PCL2 |
| Gene Symbols | MTF2 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human MTF2. The exact sequence is proprietary. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
