Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTGR1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB290747 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290747 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290747 Supplier Novus Biologicals Supplier No. NBP197748APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTGR1 Polyclonal antibody specifically detects MTGR1 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen MTGR1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias core-binding factor, runt domain, alpha subunit 2, translocated to, 2, EHTDKFZp313F2116, ETO homolog on chromosome 20, ETO homologous on chromosome 20, MTG8-like protein, MTG8-related protein 1, MTGR1p85, myeloid translocation gene-related protein 1, Myeloid translocation-related protein 1, protein CBFA2T2, ZMYND3
Host Species Rabbit
Immunogen Maps to a region between residue 554 and 604 of human Core-Binding Factor, Runt Domain, Alpha Subunit 2, translocated to, 2 using the numbering given in entry NP_005084.1 (GeneID 9139)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 9139
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.