Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTGR1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP234136 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234136 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234136 Supplier Novus Biologicals Supplier No. NBP234136
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTGR1 Polyclonal specifically detects MTGR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MTGR1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. O43439
Gene Alias core-binding factor, runt domain, alpha subunit 2; translocated to, 2, EHTDKFZp313F2116, ETO homolog on chromosome 20, ETO homologous on chromosome 20, MTG8-like protein, MTG8-related protein 1, MTGR1p85, myeloid translocation gene-related protein 1, Myeloid translocation-related protein 1, protein CBFA2T2, ZMYND3
Gene Symbols CBFA2T2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: MAKESGISLKEIQVLARQWKVGPEKRVPAMPGSPVEVKIQSRSSPPTMPPL
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 9139
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.