Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTH1 Antibody (AT3B3), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB163252 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163252 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163252 Supplier Novus Biologicals Supplier No. NBP261160G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MTH1 Monoclonal antibody specifically detects MTH1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen MTH1
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone AT3B3
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias 8-oxo-7,8-dihydrodeoxyguanosine triphosphatase, EC 3.6.1.-, MTH1Nucleoside diphosphate-linked moiety X motif 1, mutT human homolog 1,8-oxo-dGTPase, nucleoside diphosphate-linked moiety X-type motif 17,8-dihydro-8-oxoguanine triphosphatase, nudix (nucleoside diphosphate linked moiety X)-type motif 1,8-oxo-7,8-dihydroguanosine triphosphatase, Nudix motif 1
Host Species Mouse
Immunogen Recombinant human MTH1 (1-156aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Base Excision Repair, Cancer, Chromatin Research, DNA Repair, Modulation of DNA Pools, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4521
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.