Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTH1 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB355165 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355165 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355165 Supplier Novus Biologicals Supplier No. NB100109UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTH1 Polyclonal specifically detects MTH1 in Human, Mouse, Rat samples. It is validated for Gene Knock-Out, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Single Cell Western, Western Blot.

Specifications

Antigen MTH1
Applications Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Single Cell Western, Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Knockout Validated, Single Cell Western, Knockdown Validated
Formulation 50mM Sodium Borate
Gene Alias 8-oxo-7,8-dihydrodeoxyguanosine triphosphatase, EC 3.6.1.-, MTH1Nucleoside diphosphate-linked moiety X motif 1, mutT human homolog 1,8-oxo-dGTPase, nucleoside diphosphate-linked moiety X-type motif 17,8-dihydro-8-oxoguanine triphosphatase, nudix (nucleoside diphosphate linked moiety X)-type motif 1,8-oxo-7,8-dihydroguanosine triphosphatase, Nudix motif 1
Host Species Rabbit
Immunogen A C-terminal peptide derived from human MTH1, conjugated to KLH. [UniProt# P36639]
Purification Method Immunogen affinity purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Base Excision Repair, Cancer, Chromatin Research, DNA Repair, Modulation of DNA Pools, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4521
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.