Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTMR14 Antibody (OTI6B6), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N272808J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272808J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272808J549 Supplier Novus Biologicals Supplier No. NBP272808JF549

Mouse Monoclonal Antibody

MTMR14 Monoclonal specifically detects MTMR14 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MTMR14
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI6B6
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C3orf29FLJ11546, chromosome 3 open reading frame 29, EC 3.1.3.-, egg-derived tyrosine phosphatase homolog, FLJ22405, FLJ46453, FLJ90311, HCV NS5A-transactivated protein 4 splice variant A-binding protein 1, hEDTP, HJUMPY, jumpy, myotubularin related protein 14, myotubularin-related protein 14, NS5ATP4ABP1
Gene Symbols MTMR14
Host Species Mouse
Immunogen Full length recombinant protein of human MTMR14 (NP_001070993) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 64419.0
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.