Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTMR15 Antibody, Novus Biologicals™
SDP

Catalog No. NBP142677 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP142677 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP142677 Supplier Novus Biologicals Supplier No. NBP142677
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTMR15 Polyclonal specifically detects MTMR15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen MTMR15
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:100 - 1:500, Immunoprecipitation 2-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:100 - 1:500
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q9Y2M0
Gene Alias DKFZp451H236, EC 3.1.21.-, EC 3.1.4.1, FANCD2/FANCI-associated nuclease 1DKFZp686K16147, fanconi anemia associated nuclease 1, fanconi-associated nuclease 1, KIAA1018coiled-coil domain-containing protein MTMR15, MTMR15, myotubularin related protein 15, Myotubularin-related protein 15
Gene Symbols FAN1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1 and 50 of human myotubularin related protein 15 (NP_055782.3).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22909
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.