Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mtRNA polymerase Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB172064 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172064 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB172064 Supplier Novus Biologicals Supplier No. NBP318334R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

mtRNA polymerase Polyclonal antibody specifically detects mtRNA polymerase in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen mtRNA polymerase
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias APOLMTEC 2.7.7.6, DNA-directed RNA polymerase, mitochondrial, h-mtRPOL, MTRNAP, MTRPOL, polymerase (RNA) mitochondrial (DNA directed)
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 11 amino acids near the carboxyl terminus of human mtRNA polymerase. The immunogen is located within the last 50 amino acids of mtRNA polymerase.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5442
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.