Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Liofilchem™ MTS™ Vancomycin / Teicoplanin [VA/TEC] 0.5-32 / 0.5-32 μg/mL
Shop All Liofilchem Products

Click to view available options
Quantity:
10/Pk.
100/Pk.
30/Pk.
Packaging Type:
Canister Pack
Individually Packed
Description
Specifications
Specifications
| Antimicrobial Agents | Vancomycin / Teicoplanin |
| Concentration | 0.5 to 32 / 0.5 to 32 μg/mL |
| Content And Storage | -20°C or 2°C to 8°C until the given expiry date |
| Description | Liofilchem™ MTS™ Vancomycin / Teicoplanin |
| Antibiotic Code | VA/TEC |
| Product Type | MIC Test Strip |
| For Use With (Application) | Antimicrobial Resistance Detection, For determining the MIC of antimicrobial agents against microorganisms |
| Packaging Type | Canister Pack |
| Quantity | 100/Pk. |
Non-hazardous according to current legislation, but falls within the specific field of application where a safety data sheet must be supplied because it can cause phenomena of sensitization in sensitive subjects if it comes into contact with the skin.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction