Learn More
Description
Specifications
Specifications
| Antigen | MTSS1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Dilution | Western Blot, ELISA |
| Formulation | PBS |
| Gene Alias | FLJ44694, KIAA0429Missing in metastasis protein, metastasis suppressor 1, metastasis suppressor protein 1, Metastasis suppressor YGL-1, MIMA, MIMB, MIMDKFZp781P2223, MTSS1 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant MTSS1 (EAW92073.1; Met1-Ser250). |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
