Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTSS1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224716 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224716 0.1 mg
NBP224716SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224716 Supplier Novus Biologicals Supplier No. NBP224716
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

MTSS1 Polyclonal specifically detects MTSS1 in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen MTSS1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 29972794), Immunohistochemistry-Paraffin 10 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. O43312
Gene Alias FLJ44694, KIAA0429Missing in metastasis protein, metastasis suppressor 1, metastasis suppressor protein 1, Metastasis suppressor YGL-1, MIMA, MIMB, MIMDKFZp781P2223, MTSS1
Gene Symbols MTSS1
Host Species Rabbit
Immunogen A portion of amino acid 670-720 of human MTSS1 protein was used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9788
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_b), 94% homologous in monkey, 82% homologous in mouse and 70% homologous in rat.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.