Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTSS1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$192.94 - $493.10

Specifications

Antigen MTSS1
Concentration 1.0 mg/mL
Dilution Western Blot 1-4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 29972794), Immunohistochemistry-Paraffin 10 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224716SS
SDP
View Documents
Novus Biologicals
NBP224716SS
0.025 mg
Each for $192.94
Only null left
Add to Cart
 
NBP224716
SDP
View Documents
Novus Biologicals
NBP224716
0.1 mg
Each for $493.10
Only null left
Add to Cart
 
Description

Description

MTSS1 Polyclonal specifically detects MTSS1 in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

MTSS1
Western Blot 1-4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 29972794), Immunohistochemistry-Paraffin 10 ug/ml
Polyclonal
Purified
RUO
PBS with 0.05% Sodium Azide
FLJ44694, KIAA0429Missing in metastasis protein, metastasis suppressor 1, metastasis suppressor protein 1, Metastasis suppressor YGL-1, MIMA, MIMB, MIMDKFZp781P2223, MTSS1
MTSS1
IgG
Protein G purified
The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_b), 94% homologous in monkey, 82% homologous in mouse and 70% homologous in rat.
1.0 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Primate
O43312
9788
A portion of amino acid 670-720 of human MTSS1 protein was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.