Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTUS1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB166020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166020 100 μg
NB166021 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166020 Supplier Novus Biologicals Supplier No. NBP315462100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTUS1 Polyclonal antibody specifically detects MTUS1 in Human samples. It is validated for Western Blot

Specifications

Antigen MTUS1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias Angiotensin-II type 2 receptor-interacting protein, AT2 receptor-binding protein, AT2R binding protein, ATBPATIP1, ATIP, DKFZp586D1519, erythroid differentiation-related, FLJ14295, GK1, KIAA1288DKFZp686F20243, microtubule associated tumor suppressor 1, microtubule-associated tumor suppressor 1, Mitochondrial tumor suppressor 1, mitochondrial tumor suppressor gene 1, MP44, MTSG1AT2 receptor-interacting protein, transcription factor MTSG1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 207-436 of human MTUS1 (NP_065800.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer
Primary or Secondary Primary
Gene ID (Entrez) 57509
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.