Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mu Opioid R/OPRM1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11079878 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11079878 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11079878 Supplier Novus Biologicals Supplier No. NB11079878

Rabbit Polyclonal Antibody

mu Opioid R/OPRM1 Polyclonal specifically detects mu Opioid R/OPRM1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen mu Opioid R/OPRM1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P35372
Gene Alias hMOP, KIAA0403, LMOR, MOP, M-OR-1, MOR-1, MOR1MOR, Mu opiate receptor, Mu opioid receptor, mu-type opioid receptor, opioid receptor, mu 1, OPRM
Gene Symbols OPRM1
Host Species Rabbit
Immunogen Synthetic peptide comprising residues 386-400 of the human, mouse and rat MOR-1 protein.
Molecular Weight of Antigen 44.8 kDa
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4988
Test Specificity Mu Opiod Receptor
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.