Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mu Opioid R/OPRM1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP131180 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP131180 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP131180 Supplier Novus Biologicals Supplier No. NBP131180

Rabbit Polyclonal Antibody has been used in 4 publications

mu Opioid R/OPRM1 Polyclonal specifically detects mu Opioid R/OPRM1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen mu Opioid R/OPRM1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.64mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Flow Cytometry - Reported in scientific literature (PMID: 23751259)., Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence -Reported in scientific literature (PMID: 22591368)., Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS, 1% BSA, 20% Glycerol (pH7) with 0.01% Thimerosal
Gene Alias hMOP, KIAA0403, LMOR, MOP, M-OR-1, MOR-1, MOR1MOR, Mu opiate receptor, Mu opioid receptor, mu-type opioid receptor, opioid receptor, mu 1, OPRM
Gene Symbols OPRM1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human mu Opioid receptor. The exact sequence is proprietary.
Molecular Weight of Antigen 45 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4988
Test Specificity Expected to cross react based on sequence identity: Pig (86%), Rhesus Monkey (93%), Chimpanzee (86%), Bovine (86%). Not yet tested in any other species.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.