Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC1 Antibody (604804), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB376746 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376746 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376746 Supplier Novus Biologicals Supplier No. FAB6298K
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MUC1 Monoclonal antibody specifically detects MUC1 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF

Specifications

Antigen MUC1
Applications Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 604804
Conjugate DyLight 488
Dilution Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin
Host Species Mouse
Immunogen E. coli-derived recombinant human MUC-1, Pro126-Arg145, Accession # P15941
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4582
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.