Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC1 Antibody (C-Mu1), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB1002786G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002786G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002786G Supplier Novus Biologicals Supplier No. NB1002786G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MUC-1 Monoclonal specifically detects MUC-1 in Human samples. It is validated for ELISA, Immunohistochemistry, Radioimmune Assays (RIA).

Specifications

Antigen MUC-1
Applications ELISA, Immunohistochemistry, Radioimmune Assays (RIA)
Classification Monoclonal
Clone C-Mu1
Conjugate DyLight 488
Dilution ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin
Gene Symbols MUC1
Host Species Mouse
Immunogen MUC1 obtained from the breast cancer cell line T47D.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4582
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.