Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC1 Antibody (MUC1/955), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP247884A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP247884A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP247884A94 Supplier Novus Biologicals Supplier No. NBP247884AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MUC1 Monoclonal specifically detects MUC1 in Human, Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen MUC1
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone MUC1/955
Conjugate Alexa Fluor 594
Dilution Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin
Gene Symbols MUC1
Host Species Mouse
Immunogen Human milk-fat globule membranes (HMFGM) (Uniprot: P15941)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4582.0
Test Specificity This monoclonal antibody reacts with MUC1. The dominant epitope of this monoclonal antibody has not yet been determined. MUC1 is a large cell surface mucin glycoprotein expressed by most glandular and ductal epithelial cells and some hematopoietic cell lineages. It is expressed on most secretory epithelium, including mammary gland and some hematopoietic cells. It is expressed abundantly in lactating mammary glands and over expressed abundantly in 90% breast carcinomas and metastases. Transgenic MUC1 has been shown to associate with all four c-erbB receptors and localize with c-erbB1 (EGFR) in lactating glands. The MUC1 gene contains seven exons and produces several different alternatively spliced variants. The major expressed form of MUC1 uses all seven exons and is a type 1 transmembrane protein with a large extracellular tandem repeat domain. The tandem repeat domain is highly O glycosylated and alterations in glycosylation have been shown in epithelial cancer cells. Antibody to EMA is useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in bone marrow or liver.
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.