Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC1 Antibody (SM3), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N2022711PC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N2022711PC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N2022711PC7 Supplier Novus Biologicals Supplier No. NB12022711PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MUC1 Monoclonal specifically detects MUC1 in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen MUC1
Applications Flow Cytometry
Classification Monoclonal
Clone SM3
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin
Gene Symbols MUC1
Host Species Mouse
Immunogen Hydrogen fluoride deglycosylated milk mucin.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4582.0
Test Specificity Reacts very little with normal tissue. Found to react with breast, colon and ovarian carcinomas and adinocarcinoma. SM3 recognises the under-glycosylated form of MUC1 and is therefore tumour specific.
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.