Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC12 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB362313 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362313 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362313 Supplier Novus Biologicals Supplier No. NB200124MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MUC12 Polyclonal antibody specifically detects MUC12 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Radioimmune Assays

Specifications

Antigen MUC12
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Radioimmunoassay
Formulation 50mM Sodium Borate
Gene Alias MUC11, MUC-11, MUC-12, mucin 11, mucin 12, cell surface associated, mucin-11, mucin-12
Host Species Rabbit
Immunogen Synthetic peptide: WEDQNLRESRFGLEN, corresponding to amino acids 541-555 of Human MUC12.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 10071
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.