Learn More
Description
Specifications
Specifications
| Antigen | MUC12 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000 |
| Formulation | PBS and 2% Sucrose with 0.09% Sodium Azide |
| Gene Alias | MUC11, MUC-11, MUC-12, mucin 11, mucin 12, cell surface associated, mucin-11, mucin-12 |
| Gene Symbols | MUC12 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptides corresponding to MUC12(mucin 12, cell surface associated) The peptide sequence was selected from the middle region of MUC12. Peptide sequence PSVLVGDSTPSPISSGSMETTALPGSTTKPGLSEKSTTFYSSPRSPDTTH. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
