Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC13 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP224566SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224566SS 0.025 mg
NBP224566 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224566SS Supplier Novus Biologicals Supplier No. NBP224566SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MUC13 Polyclonal specifically detects MUC13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MUC13
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.25-0.50 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Alias Down-regulated in colon cancer 1FLJ20063, DRCC1, MUC-13, mucin 13, cell surface associated, mucin 13, epithelial transmembrane, mucin-13, RECC
Gene Symbols MUC13
Host Species Rabbit
Immunogen A portion of amino acids 480-512 of human Mucin 13 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56667
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.