Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC2 Antibody (994/152), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP250521FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250521FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250521FR Supplier Novus Biologicals Supplier No. NBP250521FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MUC2 Monoclonal specifically detects MUC2 in Human samples. It is validated for ELISA, Immunohistochemistry.

Specifications

Antigen MUC2
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 994/152
Conjugate DyLight 680
Dilution ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Intestinal mucin-2, MLP, MUC-2, mucin 2, intestinal/tracheal, mucin 2, oligomeric mucus/gel-forming, mucin-2, mucin-like protein, SMUC
Gene Symbols MUC2
Host Species Mouse
Immunogen This MUC2 Antibody (994/152) was developed against a peptide based on the tandem repeat sequence of MUC2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Inflammation, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4583
Test Specificity MUC2 Antibody (994/152) is specific for Muc2 and will not detect Muc3 or Muc4.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.