Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC2 Antibody (CCP58), Biotin, Novus Biologicals™
SDP

Catalog No. NBP233157B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP233157B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP233157B Supplier Novus Biologicals Supplier No. NBP233157B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MUC2 Monoclonal specifically detects MUC2 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.

Specifications

Antigen MUC2
Applications ELISA
Classification Monoclonal
Clone CCP58
Conjugate Biotin
Dilution ELISA
Gene Alias Intestinal mucin-2, MLP, MUC-2, mucin 2, intestinal/tracheal, mucin 2, oligomeric mucus/gel-forming, mucin-2, mucin-like protein, SMUC
Gene Symbols MUC2
Host Species Mouse
Immunogen A synthetic peptide of 29 amino acids, KYPTTTPISTTTMVTPTPTPTGTQTPTTT from MUC2 protein, coupled to KLH. (Uniprot: Q02817)
Molecular Weight of Antigen 520 kDa
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Inflammation, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4583
Test Specificity Recognizes a single glycoprotein of 520kDa, identified as mucin 2 (MUC2). This monoclonal antibody shows no cross-reaction with human milk fat globule membranes, MUC1, or MUC3. Its epitope has been defined as GTQTP(GlyThrGlnThrPro). Mucins are high molecular weight glycoproteins, which constitute the major component of the mucus layer that protects the gastric epithelium. MUC2 is specifically expressed in goblet cells of the small intestine in about 65% of colonic carcinomas and about 40% of gastric carcinomas. MUC2 is rarely expressed outside of the GI tract with the exceptions of mucinous carcinoma of breast and clear cell-type carcinomas of the ovary.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.