Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC2 Rabbit anti-Human, Clone: JA10-92, Novus Biologicals™
SDP

Catalog No. NBP266961 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266961 Supplier Novus Biologicals Supplier No. NBP266961
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MUC2 Monoclonal specifically detects MUC2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen MUC2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JA10-92
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias Intestinal mucin-2, MLP, MUC-2, mucin 2, intestinal/tracheal, mucin 2, oligomeric mucus/gel-forming, mucin-2, mucin-like protein, SMUC
Gene Symbols MUC2
Host Species Rabbit
Immunogen Synthetic peptide within Human MUC2 aa 5000-5049/ 5179. (SwissProt: Q02817 Human; SwissProt: Q62635 Rat)
Molecular Weight of Antigen 540 kDa
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cancer, Cellular Markers, Inflammation, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4583
Test Specificity MUC2 Antibody (JA10-91) is tissue specific for colon, small intestine, colonic tumors, bronchus, cervix and gall bladder.
Target Species Human, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.