Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC21 Rabbit anti-Human, FITC, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB182622 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB182622 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB182622 Supplier Novus Biologicals Supplier No. NBP306591F
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MUC21 Polyclonal antibody specifically detects MUC21 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen MUC21
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias bCX31G15.2, C6orf205, Chromosome 6 Open Reading Frame 205, epiglycanin, KMQK697, MUC-21, Mucin 21, Cell Surface Associated, mucin-21
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MUC21 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 394263
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.