Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC4 Antibody (781631), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB373461 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373461 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373461 Supplier Novus Biologicals Supplier No. FAB8195K
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MUC4 Monoclonal specifically detects MUC4 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen MUC4
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 781631
Conjugate DyLight 488
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Ascites sialoglycoprotein, ASGP, HSA276359, MUC-4, mucin 4, cell surface associated, mucin 4, tracheobronchial, mucin-4, Pancreatic adenocarcinoma mucin, Testis mucin, Tracheobronchial mucin
Host Species Mouse
Immunogen E. coli -derived recombinant human MUC4, Pro1072-Ser1317, Accession # Q99102
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4585
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.