Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC5AC Antibody (45M1), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB232732700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB232732700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB232732700 Supplier Novus Biologicals Supplier No. NBP232732AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MUC5AC Monoclonal specifically detects MUC5AC in Human, Mouse, Rat, Porcine, Chicken, Feline, Mammal, Monkey, Rabbit, Bovine (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot, Flow (Intracellular).

Specifications

Antigen Mucin 5AC
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 45M1
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias gastric mucin, leB, lewis B blood group antigen, major airway glycoprotein, MUC5, mucin 5, subtypes A and C, tracheobronchial/gastric, mucin 5AC, oligomeric mucus/gel-forming, mucin 5AC, oligomeric mucus/gel-forming pseudogene, mucin-5 subtype AC, tracheobronchial, mucin-5AC, TBM, tracheobronchial mucin
Gene Symbols MUC5AC
Host Species Mouse
Immunogen M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cellular Markers, Extracellular Matrix, Infections (Virus Bacteria and Parasites), Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4586
Test Specificity This monoclonal antibody recognizes the peptide core of gastric mucin M1 (recently identified as Mucin 5AC).Its epitope is located in the C-terminal cysteine rich part of the peptide core of MUC5AC. Its epitope is destroyed by beta-mercaptoethanol but not by periodate treatment. This mucin is present in primary ovarian mucinous cancer but usually absent in colorectal adenocarcinoma, thus showing an expression pattern opposite to MUC2. Together with a panel of antibodies, Anti-MUC5AC may be useful for differential identification of primary mucinous ovarian tumors from colon adenocarcinoma metastatic to the ovary. MUC5AC antibodies may also be useful for identification of intestinal metaplasia as well as in the identification of pancreatic carcinoma and pre-cancerous changes vs. normal pancreas.
Target Species Human, Mouse, Rat, Pig, Chicken, Feline, Primate, Rabbit, Bovine (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.