Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC5AC Antibody (MUC5AC/7066R), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB178029 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB178029 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB178029 Supplier Novus Biologicals Supplier No. NBP314003AF488
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MUC5AC Monoclonal antibody specifically detects MUC5AC in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen MUC5AC
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MUC5AC/7066R
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias gastric mucin, leB, lewis B blood group antigen, major airway glycoprotein, MUC5, mucin 5, subtypes A and C, tracheobronchial/gastric, mucin 5AC, oligomeric mucus/gel-forming, mucin 5AC, oligomeric mucus/gel-forming pseudogene, mucin-5 subtype AC, tracheobronchial, mucin-5AC, TBM, tracheobronchial mucin
Host Species Rabbit
Immunogen Recombinant full-length human MUC5AC protein (Uniprot: P98088 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Extracellular Matrix, Infections (Virus Bacteria and Parasites), Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4586
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.