Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC5B Antibody (4A10-H2), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP250522AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250522AF 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250522AF Supplier Novus Biologicals Supplier No. NBP250522AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MUC5B Monoclonal specifically detects MUC5B in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Mucin 5B
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 4A10-H2
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias high molecular weight salivary mucin MG1, mucin 5, subtype B, tracheobronchial, mucin 5B, oligomeric mucus/gel-forming, sublingual gland mucin, tracheobronchial
Gene Symbols MUC5B
Host Species Mouse
Immunogen KLH conjugated peptide RNREQVGKF in 4 of the cysteine domains of the tandem repeat region was used to immunise Balb/c female mice.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Cellular Signaling
Primary or Secondary Primary
Gene ID (Entrez) 727897
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.