Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC6 Antibody (MUC6/7069R), FITC, Novus Biologicals™
SDP

Catalog No. NB178084 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB178084 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB178084 Supplier Novus Biologicals Supplier No. NBP314010F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MUC6 Monoclonal antibody specifically detects MUC6 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen MUC6
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MUC6/7069R
Conjugate FITC
Formulation PBS
Gene Alias gastric mucin, Gastric mucin-6, MUC-6, mucin 6, gastric, mucin 6, oligomeric mucus/gel-forming, mucin-6
Host Species Rabbit
Immunogen Recombinant human MUC6 protein (Uniprot: Q6W4X9 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4588
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.