Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC7 Antibody (4D2-1D7), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP250391V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250391V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250391V Supplier Novus Biologicals Supplier No. NBP250391V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MUC7 Monoclonal specifically detects MUC7 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen MUC7
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 4D2-1D7
Conjugate DyLight 405
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias apo-MG2, DKFZp686J03256, FLJ27047, MG2MGC34772, MUC-7, mucin 7, secreted, mucin-7, salivary, Salivary mucin-7
Gene Symbols MUC7
Host Species Mouse
Immunogen KLH conjugated peptide EGRERDHELRHRRHHHQ located in the N-terminal part of the MUC7 protein within the histatin-like domain.was used to immunise Balb/c female mice.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4589
Test Specificity No cross reactivity with other human mucins.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.