Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Mucin 6 (MUC-6) Ab-1, Mouse Monoclonal Antibody

Deliver accurate, reproducible results with the Epredia™ Mucin 6 (MUC-6) Ab-1, Mouse Monoclonal Antibody.

Specifications

Antigen Mucin 6 (MUC-6) Ab-1
Clone CLH5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
MS1153S0
View Documents
Epredia™
MS1153S0
Concentrated 100 μL N/A N/A N/A
MS1153S1
View Documents
Epredia™
MS1153S1
Concentrated 500 μL N/A N/A N/A
MS1153S
View Documents
Epredia™
MS1153S
Concentrated 1 mL N/A N/A N/A
MS1153R7
View Documents
Epredia™
MS1153R7
Prediluted 7 mL N/A N/A N/A
Description

Description

Mucin 6 (Muc-6 Glycoprotein). Mucins are heavily glycosylated glycoproteins, which constitute the major components of mucus which cover the surface of epithelial tissues. Nine distinct epithelial mucin genes (Muc-1, 2, 3, 4, 5AC, 5B, 6, 7 and 8) have been identified. Various immunohistochemical and in situ hybridization studies have shown that these mucins are differentially expressed in epithelia with cell-type specificity. Muc-5AC and Muc-6 glycoproteins are the first two mucins found in superficial epithelium and Muc-6 glycoprotein is found in the deep glands.

Host Species: Mouse

Clone: CLH5

Isotype: IgG1

Species Reactivity: Human. Weakly react with Rat. Others not known.

Epitope: Not determined

Immunogen: Synthetic peptide of the tandem repeat sequence of human MUC-6 protein

Positive Control: Stomach

Cellular Localization: Cytoplasmic and cell surface

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Mucin 6 (MUC-6) Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
IgG1
Stomach
CLH5
Monoclonal
Mouse
Cancer and Tumor Biology
Synthetic peptide of the tandem repeat sequence of human Muc-6 protein
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.