Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mucin13 Mouse anti-Human, Unconjugated, Clone: C14, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC1133100UL
Description
Mucin13 Monoclonal antibody specifically detects Mucin13 in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Inhibits Activity/Function, Western Blotting.
Specifications
Mucin13 | |
Monoclonal | |
Unconjugated | |
MUC-13 | |
Mouse | |
54.60 kDa calculated. | |
100 μL | |
Primary | |
Human | |
Purified |
ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
C14 | |
NP_149038 | |
MUC13, DRCC1, RECC, UNQ6194/PRO20221 | |
GST-tagged full-length human recombinant Mucin-13. | |
Protein G Purified | |
Signaling | |
Clone C14 is a mouse monoclonal antibody that specifically detects human Mucin-13. | |
Stable for 1 year at -20°C from date of receipt. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction