Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mucin13 Mouse anti-Human, Unconjugated, Clone: C14, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $457.74
Specifications
Antigen | Mucin13 |
---|---|
Clone | C14 |
Applications | ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABC11332MI
|
MilliporeSigma
MABC113325UL |
25 μL |
Each for $182.31
|
|
|||||
MABC11331MI
|
MilliporeSigma
MABC1133100UL |
100 μL |
Each for $457.74
|
|
|||||
Description
Mucin13 Monoclonal antibody specifically detects Mucin13 in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Inhibits Activity/Function, Western Blotting.Specifications
Mucin13 | |
ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
MUC-13 | |
GST-tagged full-length human recombinant Mucin-13. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
54.60 kDa calculated. |
C14 | |
Monoclonal | |
Purified | |
Signaling | |
NP_149038 | |
MUC13, DRCC1, RECC, UNQ6194/PRO20221 | |
IgG κ | |
Protein G Purified | |
Clone C14 is a mouse monoclonal antibody that specifically detects human Mucin-13. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title