Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mucolipin 1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB339004 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339004 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339004 Supplier Novus Biologicals Supplier No. NB11082375MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mucolipin 1 Polyclonal antibody specifically detects Mucolipin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot

Specifications

Antigen Mucolipin 1
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Simple Western
Formulation 50mM Sodium Borate
Gene Alias MG-2, ML4TRP-ML1, MLIV, MST080, MSTP080, mucolipidin, mucolipidosis type IV protein, mucolipin 1, mucolipin-1, TRPML1, TRP-ML1, TRPM-L1
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of mouse Mucolipin 1 (within residues 500-580). [Swiss-Prot# Q99J21]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Neuroscience, Sensory Systems, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 57192
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.