Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUL1 Antibody (3G2.2F7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP23136201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP23136201 0.1 mg
NBP23136225 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP23136201 Supplier Novus Biologicals Supplier No. NBP2313620.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MUL1 Monoclonal specifically detects MUL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MUL1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3G2.2F7
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q969V5
Gene Alias C1orf166mitochondrial E3 ubiquitin ligase 1, E3 ubiquitin-protein ligase MUL1, EC 6.3.2, EC 6.3.2.-, FLJ12875, GIDERP11-401M16.2, Growth inhibition and death E3 ligase, MAPLE3 ubiquitin ligase, mitochondrial E3 ubiquitin protein ligase 1, mitochondrial ubiquitin ligase activator of NF-kB, mitochondrial ubiquitin ligase activator of NFKB 1, Mitochondrial-anchored protein ligase, MULANchromosome 1 open reading frame 166, Putative NF-kappa-B-activating protein 266, RING finger protein 218, RNF218mitochondria-anchored protein ligase
Gene Symbols MUL1
Host Species Mouse
Immunogen Partial recombinant protein made to a C-terminal portion of the human MUL1 protein (between residues 200-352) [UniProt Q969V5]
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 79594
Test Specificity The immunogen sequence shows 99% similarity to Monkey's MUL1 and is 84% similar to isoform 1 and 3 of Mouse's MUL1.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.