Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUL1 Antibody (3G2.2F7), PE, Novus Biologicals™
SDP

Catalog No. NB337066 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337066 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337066 Supplier Novus Biologicals Supplier No. NBP231362PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MUL1 Monoclonal antibody specifically detects MUL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen MUL1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3G2.2F7
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias C1orf166mitochondrial E3 ubiquitin ligase 1, E3 ubiquitin-protein ligase MUL1, EC 6.3.2, EC 6.3.2.-, FLJ12875, GIDERP11-401M16.2, Growth inhibition and death E3 ligase, MAPLE3 ubiquitin ligase, mitochondrial E3 ubiquitin protein ligase 1, mitochondrial ubiquitin ligase activator of NF-kB, mitochondrial ubiquitin ligase activator of NFKB 1, Mitochondrial-anchored protein ligase, MULANchromosome 1 open reading frame 166, Putative NF-kappa-B-activating protein 266, RING finger protein 218, RNF218mitochondria-anchored protein ligase
Host Species Mouse
Immunogen Partial recombinant protein made to a C-terminal portion of the human MUL1 protein (between residues 200-352) [UniProt Q969V5]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 79594
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.