Learn More
Multimedia Packages: Mitosis and Meiosis
SCI_ED
Catalog No. S90071A Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Type:
Student Package
Teacher Package
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S90071A Student Package
S90071 Teacher Package
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S90071A Supplier Johannes Lieder GmbH & Co. KG Supplier No. SSE21
Only null left
Add to Cart
Add to Cart
Includes
Teacher Package:
  • Six microscope slides in plastic box
  • Three overhead projector color transparencies
  • Six sketch- and worksheets
  • Brochure with explanatory text
  • Cardboard storage box
Student Package:
  • Six microscope slides in plastic box
  • Brochure with explanatory text
  • Cardboard storage box
Six selected microscope slides with accompanying brochure.
Teacher and Student Packages, set of six slides:
  • Allium, root tips, l.s. showing lateral view of all stages of mitosis, iron-hematoxyline
  • Whitefish mitosis, l.s. of embryo showing animal mitosis
  • Testis of mouse, t.s. showing spermatogenesis in all stages
  • Giant chromosomes from salivary gland of Chironomus, squash preparation special stained for chromomeres
  • Lilium, microspore mother cells, prophase of first division showing meiosis
  • Lilium, microspore mother cells, meta- or anaphase of first division, showing mitosis
Specifications
Type Student Package
Quantity 6