Learn More
Millipore Multiscreen™ 96-Well Plate, Polystyrene, Solid Bottom
Catalog No. p-200080423 Shop All MilliporeSigma Products
Change view
Click to view available options
Color:
Black
Clear
White
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round Bottom (U-well)
Catalog No. Color Well Shape
MSSBNFX40 Black Flat Bottom (F-well)
MSSCNFX00 Clear Flat Bottom (F-well)
MSSCNUX00 Clear Round Bottom (U-well)
MSSCNVX00 Clear Conical Bottom (V-well)
MSSWNFX40 White Flat Bottom (F-well)
5 options
Catalog No. MSSBNFX40
Supplier: Millipore MSSBNFX40
Only null left
Add to Cart
Add to Cart
Non-sterile polystyrene plates for use in common biochemical assay platforms.
  • MultiScreen™ 96-well polystyrene microplates meet industry standard dimensions for use in common biochemical assay platforms, including automated liquid handling, microplate readers, and other microplate compatible devices.
  • Polystyrene microplates are appropriate for a variety of biochemical assays in addition to sample preparation and storage.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Array 8 x 12
Color Black
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 300 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)